Analytical standard, Sulfapyridine-phenyl-¹³C₆, reference material, Honeywell Fluka™
Catalog # HONCR1017-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-45-3
- Description:Sulfapyridine-phenyl-¹³C₆
- Melting point:180…184 °C
- Formula:C₅¹³C₆H₁₁N₃O₂S
- Storage temperature:Ambient
- Description:Sulfapyridine-phenyl-¹³C₆
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results