Order Entry
Northern Ireland
ContactUsLinkComponent
 
Analytical standard, Hydroxymethylclenbuterol hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1007-10MG
CAS Number:  
Analytical standard, Hydroxymethylclenbuterol hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1007-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    37162-89-3
  • Description:
    Hydroxymethylclenbuterol hydrochloride
  • Melting point:
    215…216 °C
  • Formula:
    C₁₂H₁₈Cl₂N₂O₂ · HCl
  • Storage temperature:
    2…8 °C
  • Description:
    Hydroxymethylclenbuterol hydrochloride

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results