Order Entry
Northern Ireland
ContactUsLinkComponent
 
Analytical standard, Dapsone-¹³C₁₂, reference material, Honeywell Fluka™
  HONCR1004-10MG
 :  
Analytical standard, Dapsone-¹³C₁₂, reference material, Honeywell Fluka™
  HONCR1004-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1632119-29-9
  • Description:
    Dapsone-¹³C₁₂
  • Melting point:
    174 °C
  • Formula:
    ¹³C₁₂H₁₂N₂O₂S
  • Storage temperature:
    Ambient
  • Description:
    Dapsone-¹³C₁₂

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results