Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
L(+)-Tartaric acid 99.7-100.5% (dried basis), fine-granular NF, FCC, Macron Fine Chemicals™
  MACR2307-04
 :  
undefined
 
L(+)-Tartaric acid 99.7-100.5% (dried basis), fine-granular NF, FCC, Macron Fine Chemicals™
  MACR2307-04
 :  

 

  • Pack type:
    PE bottle, Mall Polystormor®
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Bulk Food Chemical:
    99.7 - 100.5%
  • NF - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry):
    99.7 - 100.5 %
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Loss on Drying:
    ≤0.5 %
  • NF - Residue on Ignition:
    ≤0.1 %
  • NF - Limit of Oxalate:
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Optical Rotation [Å]²⁵^D (+):
    12.2 - 13.0 Degree
  • FCC - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry):
    99.7 - 100.5 %
  • FCC - Identification A (IR):
    Passes Test
  • FCC - Identification B (Tartrate):
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Limit of Oxalate:
    Passes Test
  • FCC - Loss on Drying:
    ≤0.5 %
  • FCC - Residue on Ignition:
    ≤0.05 %
  • FCC - Specific (Optical) Rotation [Å]²⁵^D (+):
    12.2 - 13.0 Degree
  • FCC - Sulfate (SO₄):
    Passes Test
  • Solubility (20 in 100):
    Passes Test

 

 

 :
 :13,09158
 :399 °C (1013 hPa)
 :206 °C
 :1,759 g/cm³ (20 °C)
 :> 100 °C (closed cup)
 :150.09 g/mol
 :Ambient
 :201-766-0
 :01-2119537204-47
 :GHS05
 :87-69-4
 :C₄H₆O₆
 :MFCD00064207