Eye shower, wall mounted
Catalog # BROE17550009
Supplier: BROEN-LAB
Specifications
- Description:Eye shower, wall mounted, 4,5 kg
- Pk:1
Specifications
About this item
Body and eye shower, wall mounted in brass with chemical resistant BROEN-LAB Polycoat with self-draining shower head.
- Reliable and efficient showers for eyes and face
- Professional solutions for employee safety
- Easy to mount
- Anti-limescale design and material
- Improved functionality with long lifetime
- Strong wall flange with 6 holes for wall mounted models with exposed piping
The water amount is regulated by a built-in FLOWFIX (14 L/min).
Min. working pressure: 1,5 bar.
Certifications: EN 15154-1; EN 15154-2