Indoor Outdoor Vinyl Labels, M211 and M210
Supplier: Brady
Certificates
About this item
These B-595 vinyl labels will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
- Average outdoor durability: 8 to 10 years
- Gloss finish
- For smooth and textured/rough surfaces
The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
Frequently Bought Together