ContactUsLinkComponent
pET-30a(+) DNA, Novagen®
pET-30a(+) DNA, Novagen®
Catalog # 69909-3
CAS Number:  
pET-30a(+) DNA, Novagen®
Catalog # 69909-3
CAS Number:  
Specifications
  • Pk:
    10 µG
  • Description:
    pET-30a(+) DNA, Novagen®
Specifications
About this item
The pET-30a-c(+) vectors carry an N-terminal His Tag®/thrombin/S Tag™/enterokinase configuration plus an optional C-terminal His tag sequence.
Delivery: The pET Vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).