

Ergocalciferol (vitamin D2, calciferol) 97.0-103.0%
Catalog # J15562.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Assay (unspecified):97.0 - 103.0% (as is)
- Assay (unspecified - Reducing Substances):Passes Test
- Identification (A, B, C, D):Passes Test
- Optical Rotation (c= 1.5 in EtOH):+103 to 106°
Specification Test Results
About this item
:
Merck Index:14,10018
Melting Pt:118…119 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,II
:396.66 g/mol
EINECS:200-014-9
:GHS06
:GHS08
CAS Number:50-14-6
Formula:C₂₈H₄₄O
MDL Number:MFCD00166988