Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ninhydrin reagent kit
Ninhydrin reagent kit L8500
Ninhydrin reagent kit L8500
Ninhydrin reagent kit L8500
Catalog # 299-70501
Supplier:
VWR Chemicals
CAS Number:
Certificates
Expand image
Ninhydrin reagent kit L8500
Catalog # 299-70501
Supplier:
VWR Chemicals
CAS Number:
Certificates
Specifications
Pk:
1 L
Seq:
5037
Pk Info:
1 set×2 l
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation