
Sodium hydroxide 1 mol/l (1N), pharma grade
APPC192415.1212
:
- Pk:2,5 L
- Seq:5096
- Titer at 20 °C:1.00 ±0.01
- Identity According to Pharmacopoeias:Passes Test
- Chloride (Cl):Max. 0.005%
- Nitrogen compounds (as N):Max. 0.002%
- Phosphate (PO₄):Max. 0.002
- Sulfate (SO₄):Max. 0.005%
- Carbonate (as Na₂CO₃):Max. 1.0%
- Residual solvents (Ph. Eur./USP):Passes Test
- Heavy metals (as Pb):Max. 0.001%
- Al:Max. 0.002%
- Ca:Max. 0.005%
- Fe:Max. 0.001%
- K:Passes Test
- MG:Max. 0.005%
- Cd, Class 1 - (ICH Q3D):Max. 0.5 ppm
- Pb, Class 1 - (ICH Q3D):Max. 5 ppm
- As, Class 1 - (ICH Q3D):Max. 0.4 ppm
- Hg, Class 1 - (ICH Q3D):Max. 0.1 ppm
- Co, Class 2A - (ICH Q3D):Max. 5 ppm
- V, Class 2A - (ICH Q3D):Max. 10 ppm
- Ni, Class 2A - (ICH Q3D):Max. 10 ppm
- Tl, Class 2B - (ICH Q3D):Max. 5 ppm
- Au, Class 2B - (ICH Q3D):Max. 10 ppm
- Pd, Class 2B - (ICH Q3D):Max. 10 ppm
- Ir, Class 2B - (ICH Q3D):Max. 10 ppm
- Os, Class 2B - (ICH Q3D):Max. 10 ppm
- Rh, Class 2B - (ICH Q3D):Max. 10 ppm
- Ru, Class 2B - (ICH Q3D):Max. 10 ppm
- Se, Class 2B - (ICH Q3D):Max. 15 ppm
- Ag, Class 2B - (ICH Q3D):Max. 15 ppm
- Pt, Class 2B - (ICH Q3D):Max. 10 ppm
- Li, Class 3 - (ICH Q3D):Max. 55 ppm
- Sb, Class 3 - (ICH Q3D):Max. 120 ppm
- Ba, Class 3 - (ICH Q3D):Max. 140 ppm
- Mo, Class 3 - (ICH Q3D):Max. 25 ppm
- Cu, Class 3 - (ICH Q3D):Max. 10 ppm
- Sn, Class 3 - (ICH Q3D):Max. 600 ppm
- Cr, Class 3 - (ICH Q3D):Max. 25 ppm
:
:40 g/mol
:1,042 g/cm³
:Ambient
:215-185-5
:1824
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548