ESD safety ankle boots, lace-up, Maverick Black Mid
Supplier: ELTEN
Certificates
About this item
Leather-free ESD safety ankle boots made from hydrophonised microfibre and hydrophobised textile material, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU sole (Fun).
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (ESD Pro), black
- Anatomically-shaped insole and ankle support for improved stability
- Protective steel toecap (200 J)
- Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black or black/red
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- 114-8979
- 114-8978
- 114-8977
- 114-8976
- 114-8975
- 114-8974
- 114-8973
- 114-8972
- 114-8971
- 114-8970
- 114-8969
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001