ESD safety ankle boots, lace-up, L10, Emotion
Supplier: ELTEN
Certificates
About this item
Lightweight, vintage and casual style, environmental friendly manufactured ESD safety ankle boot made from hydrophobised nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
- Fabric lining Bioactive
- Replaceable insole; orthopaedic insoles possible
- Full-length inlay sole ESD PRO beige
- Metal-free puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Beige
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- 114-8816
- 114-8822
- 114-8815
- 114-8821
- 114-8814
- 114-8820
- 114-8819
- 114-8813
- 114-8817
- 114-8812
- 114-8811
- 114-8818
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001