ContactUsLinkComponent
Positive electrophoretic mobility standard, NIST-1980
Catalog # PROONIST-1980
CAS Number:  
Positive electrophoretic mobility standard, NIST-1980
Catalog # PROONIST-1980
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Electrophoretic mobility standard
Specifications
About this item
This Standard Reference Material (SRM) is intended for use in the evaluation of equipment, and the validation of methodology used to measure electrophoretic mobility.
  • Packaging of 500 mg/L
A unit of SRM 1980 consists of a single 60 cm³ polyethylene bottle containing 40 cm³ of suspension, which is to be diluted prior to analysis. The suspension consists of 500 mg/l goethite (a-FeOOH) suspension saturated with 100 μmol/g phosphate in a 5×10⁻² mol/L sodium perchlorate electrolyte solution at a pH of 2,5.
The goethite powder consists of acicular particles with an average dimension of 60×20 nm as determined from electron microscopy.
Certifications: NIST, SRM Certified.