ContactUsLinkComponent
Combined shower and eyewash test kit
Combined shower and eyewash test kit
Catalog # SPEASE-950
Supplier:  SPEAKMAN
Combined shower and eyewash test kit
Catalog # SPEASE-950
Supplier:  SPEAKMAN
Specifications
  • Description:
    Combined shower and eyewash test kit
  • Pk:
    1
Specifications
About this item
The shower and eyewash test kit is designed for testing of emergency safety showers and eyewashes to validate that the equipment meets ANSI/ISEA Z358.1 compliance.
  • Features vinyl funnel sleeve with PVC handle
Delivery: Includes 18,9 l bucket and lid, shower tester, and eyewash tester.