PIG® Fat Mat® super-absorbent mat
Catalog # NEWPMAT2101
Supplier: NEW PIG
Specifications
- Description:Super-absorbent mat pads, extremeweight
- L×W:510×410 mm
- Absorbency:83,3 L
- Packed:50 pads/dispenser
- Pk:50
Specifications
About this item
This double-thick version of 8-layer PIG Mat has 16 thirsty layers for big cleanups. Spunbond top resists abrasion for long life.
- 16 layers of 100% polypropylene are thermally bonded to form one super-absorbent mat capable of drinking up huge volumes of liquid
- Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated
- Fast-wicking 'diaper action' transfers liquids into the absorbent inner layers to keep moisture away from surface
- Absorbs and retains oils, coolants, solvents and water
- Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
- Can be wrung out and incinerated after use to reduce waste or for fuels blending
- Dispenser box makes it easy to take, carry and store your mat whilst protecting it from dirt and moisture
Easy-tear perforations let you take only what you need, so you use less mat and save money. Extreme-weight Fat Mat construction is ideal for applications that require long life and high capacity.