VWR® Mollifex™, Softening agent
Catalog # 360584X
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:Softening agent Mollifex™
- Seq:01
- M-Clarity quality level:MQ100
Specifications
About this item
Mollifex™ is used to soak the cut face of paraffin embedded tissue blocks prior to sectioning is useful to facilitate the mechanical process of sectioning.
- Flash point 20 °C
- Store at 15 to 30 °C
By rehydrating the tissue, Mollifex allows it to be cut smoothly rather than be fractured. This process also reduces the common problems of uneven staining and loss of detail which result from improper or excessive processing of some tissues.