4-Methyl-3-thiosemicarbazide ≥97%
Catalog # B21138.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:105.16 g/mol
Boiling Pt:201 °C (1013 hPa)
Melting Pt:136 °C
Density:1,362 g/cm³ (20 °C)
EINECS:229-563-2
UN:2811
ADR:6.1,II
CAS Number:6610-29-3
Formula:C₂H₇N₃S
MDL Number:MFCD00007617