
2-Acetylthiophene
Catalog # MOLEM77687659
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:126.18 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:9 °C
Density:1,1704 g/cm³ (20 °C)
EINECS:201-804-6
UN:2810
ADR:6.1,II
:GHS06
CAS Number:88-15-3
Formula:C₆H₆OS
MDL Number:MFCD00005442