Order Entry
Northern Ireland
ContactUsLinkComponent
2-Acetylthiophene for synthesis, Sigma-Aldrich®
2-Acetylthiophene for synthesis, Sigma-Aldrich®
Catalog # 8.00175.0250
Supplier:  Merck
CAS Number:  
2-Acetylthiophene for synthesis, Sigma-Aldrich®
Catalog # 8.00175.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    14 g/l (30 °C)
  • Melting Point:
    9 - 11 °C
  • Molar Mass:
    126.18 g/mol
  • Boiling Point:
    211 - 213 °C (1013 hPa)
  • Flash Point:
    97 °C
  • Density:
    1.17 g/cm3 (20 °C)
  • Ignition Point:
    265 °C DIN 51794

Specification Test Results

About this item

 :
 :126.18 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:9 °C
Density:1,1704 g/cm³ (20 °C)
EINECS:201-804-6
UN:2810
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS06
CAS Number:88-15-3
Formula:C₆H₆OS
MDL Number:MFCD00005442