2,4-Diamino-6-hydroxypyrimidine for synthesis
Catalog # 8.41789.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Melting Point:285 - 286 °C
- Molar Mass:126.11 g/mol
- Hill Formula:C4H6N4O
Specification Test Results
About this item
:
:126.12 g/mol
Merck Index:13,03001
Boiling Pt:373 °C (1013 hPa)
Melting Pt:285…286 °C
:M-Clarity™ quality level = MQ200
EINECS:200-254-4
CAS Number:56-06-4
:MFCD08528919
Formula:C₄H₆N₄O
MDL Number:MFCD00006098