2,4-Diamino-6-hydroxypyrimidine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-06-4
:MFCD08528919
Formula:C₄H₆N₄O
MDL Number:MFCD00006098
:126.12 g/mol
Merck Index:14,2981
Boiling Pt:373 °C (1013 hPa)
Melting Pt:285…286 °C (dec.)
Storage Temperature:Ambient
EINECS:200-254-4
Related Information
Frequently Bought Together