2,4-Dichloropyrimidine
Glass bottle for solids, 25 g
Catalog # MOLEM17839790
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:3934-20-1
- Formula:C₄H₂Cl₂N₂
- MDL Number:MFCD00006061
- Molecular Weight:148.98 g/mol
- Boiling Pt:209 °C (1013 hPa)
- Melting Pt:61 °C
- EINECS:223-508-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



