
2-Amino-4-methyl-3-nitropyridine 98%
Catalog # ACRO319550250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:136 to 141 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:153.14 g/mol
Melting Pt:136…141 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:6635-86-5
Formula:C₆H₇N₃O₂
MDL Number:MFCD00006315