2-Amino-3-iodo-5-methylpyridine 95%
Catalog # H27787.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:211308-79-1
:
Formula:C₆H₇IN₂
:234.04 g/mol
Melting Pt:86…89 °C
:GHS06
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD09037445