Order Entry
Northern Ireland
ContactUsLinkComponent
5-Bromo-2-methylpyridine ≥98%
5-Bromo-2-methylpyridine ≥98%
Catalog # H27085.06
CAS Number:  
5-Bromo-2-methylpyridine ≥98%
Catalog # H27085.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :172.02 g/mol
Melting Pt:32…36 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:3430-13-5
Formula:C₆H₆BrN
MDL Number:MFCD00661170