Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Rat Recombinant LEPTIN (from <i>E. coli</i>)

Rat Recombinant LEPTIN (from E. coli)

Vial, 200 µG
Catalog # SHBT300-14-200UG
Rat Recombinant LEPTIN (from <i>E. coli</i>)
Catalog # SHBT300-14-200UG
CAS Number:  
Rat Recombinant LEPTIN (from <i>E. coli</i>)
Rat Recombinant LEPTIN (from E. coli)

Vial, 200 µG

CAS Number:  
Price
£65.20
Log in for availability
1 * 200 µG
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Macrophage inflammatory protein-1 alpha (MIP-1 α), also known as CCL3, is a cytokine produced by macrophages. MIP-1 α binds the chemokine receptors CCR1, CCR4 and CCR5 to induce inflammatory responses, including the recruitment of granulocytes and neutrophil superoxide production. The MIP-1 α and MIP-1 β heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1).

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 µG
  • Protease-free:
    N
  • UniProtKB:
    P50596
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    16,3 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Protein function:
    Cytokine
  • Gene ID:
    P50596
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Endotoxin-free:
    N
  • Species:
    Rat
  • Protein/peptide name:
    Leptin
  • Protein synonyms:
    Obesity protein
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT300-14-200UG
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Storage conditions:
    −20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.