Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-3-iodopyridine
2-Fluoro-3-iodopyridine ≥97%
2-Fluoro-3-iodopyridine ≥97%
2-Fluoro-3-iodopyridine ≥97%
H50146.03
:
Thermo Fisher Scientific
:
2-Fluoro-3-iodopyridine ≥97%
H50146.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
113975-22-7
:
:
C₅H₃FIN
:
MFCD03095287
:
222.99 g/mol
:
GHS07
:
Ambient