
Pyridine-2,3-dicarboxylic anhydride
Catalog # MOLEM26885425
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:149.11 g/mol
Boiling Pt:360 °C (1013 hPa)
Boiling Pt:360 °C (1013 hPa)
Melting Pt:146 °C
EINECS:211-834-1
:GHS07
CAS Number:699-98-9
Formula:C₇H₃NO₃
MDL Number:MFCD00005915