Order Entry
Northern Ireland
ContactUsLinkComponent
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Catalog # 47864.
Supplier:  Merck
CAS Number:  
undefined
Nicotinic acid (Niacin)
Catalog # 47864.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3
 :
 :123.11 g/mol
Boiling Pt:258
EINECS:200-441-0
 :GHS07
Melting Pt:142
Storage Temperature:Ambient
CAS Number:59-67-6
Formula:C₆H₅NO₂
MDL Number:MFCD00006391
Density:1.473 g/cm³ (20 °C)
Flash Pt:193
Merck Index:13,06552