2-Chloro-4-methoxypyridine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17228-69-2
:
Formula:C₆H₆ClNO
MDL Number:MFCD02093951
:143.57 g/mol
Boiling Pt:110…115 °C (10 mmHg)
Density:1,258
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together