
5-Bromo-2-methoxypyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13472-85-0
:
Formula:C₆H₆BrNO
MDL Number:MFCD01318952
Boiling Pt:80 °C (12 mmHg)
Density:1,453
Flash Pt:96 °C (204 °F)
:188.02 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together