2-Bromo-5-chloropyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40473-01-6
:
Formula:C₅H₃BrClN
MDL Number:MFCD00234006
:192.44 g/mol
Melting Pt:65…69 °C
UN:2811
:GHS07
Specification Test Results
- Appearance:Beige to yellow-brown Powder
- Infrared spectrum:Conforms
- Melting point:65 to 69 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together