
3,6-Dichloropyridazine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-30-0
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00006466
:148.98 g/mol
Boiling Pt:89…91 °C (0,2 mmHg)
Melting Pt:70 °C
Density:1,7 g/cm³ (20 °C)
EINECS:205-478-6
ADR:6.1,II
:M-Clarity™ quality level = MQ200
UN:2811
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.0012 g/l (20 °C)
- Melting Point:72 °C
- Molar Mass:148.98 g/mol
Specification Test Results
Frequently Bought Together