
Pyrazine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:290-37-9
:
Formula:C₄H₄N₂
MDL Number:MFCD00006122
:80.09 g/mol
Merck Index:13,08047
Boiling Pt:115 °C (1013 hPa)
Melting Pt:52 °C
Density:1,0311 g/cm³ (60.9 °C)
UN:1325
ADR:4.1,II
:M-Clarity™ quality level = MQ200
EINECS:206-027-6
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:49 - 52 °C
- Molar Mass:80.08 g/mol
- Boiling Point:115 - 118 °C (1013 hPa)
- Synonyms:1,4-Diazine
- Hill Formula:C4H4N2
Specification Test Results
Frequently Bought Together