
2-Aminopurine
Catalog # MOLE57189043-100MG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:135.13 g/mol
Melting Pt:277…278 °C
EINECS:207-197-4
:GHS07
Storage Temperature:Refrigerator
CAS Number:452-06-2
Formula:C₅H₅N₅
MDL Number:MFCD00005566