Order Entry
Northern Ireland
ContactUsLinkComponent
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
Irritant
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
Catalog # MOLE10920590-25G
Supplier:  Molekula
CAS Number:  
Irritant
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
Catalog # MOLE10920590-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:351 °C (1013 hPa)
Melting Pt:–20 °C
Density:1,11 g/cm³ (20 °C)
 :220.27 g/mol
Storage Temperature:Ambient
EINECS:251-379-6
 :GHS07
CAS Number:33100-27-5
Formula:C₁₀H₂₀O₅
MDL Number:MFCD00005110