About this item
Interleukin 17AF (IL-17AF) is a heterodimer that is composed of the interleukin 17A (IL-17A) and interleukin 17F (IL-17F) members of the IL-17 family of cytokines. IL-17AF is produced by T helper 17 cells (Th17) following interleukin 23 (IL-23) stimulation. IL-17AF signals through the IL-17RA/IL-17RC receptor complex and functions to regulate inflammatory responses. IL-17AF induces chemokine and airway neutrophilia production, similar in function to IL-17A and IL-17F homodimers. In regard to these functions, IL-17AF is less active than the IL-17A homodimer and shows greater activity than the IL-17F homodimer. Human and rat IL-17AF are active on mouse cells.
- High quality
- Low endotoxin
- Affordable
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: Production of IL-6 from mouse 3T3 cells, ED₅₀ ≤50 ng/ml, Bioactivity ≥2,0×10⁴ units/mg
- Gene ID:Q16552/Q96PD4
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:IL-17AF
- Purity:≥95%
- Molecular weight:30,7 kDa
- Sequence:IL-17A:MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA IL-17F:MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay