
Dimidium bromide, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:518-67-2
:
Formula:C₂₀H₁₈BrN₃
MDL Number:MFCD00011757
:380.29 g/mol
Melting Pt:243…248 °C
Storage Temperature:Ambient
EINECS:208-256-7
UN:2811
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:242 °C
- Molar Mass:380.28 g/mol
- Bulk Density:380 kg/m3
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Frequently Bought Together