Order Entry
Northern Ireland
ContactUsLinkComponent
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Appearance of solution:
    passes test
  • Copper (Cu):
    max. 5 ppm
  • Iron (Fe):
    max. 2 ppm
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Oxalic acid:
    passes test
  • Specific Optical Rotation:
    +20.5 - +21.5 °
  • Sulfated Ash:
    max. 0.10 %

Specification Test Results

Related Information

Frequently Bought Together