CD40 ligand (CD40L) or CD154 is a membrane glycoprotein and differentiation antigen that is expressed on the surface of T cells, monocytes, basophils, eosinophils, platelets, dendritic cells, and endothelial cells.
- Produced from genetically modified E. coli
- Monomeric protein, containing 149 amino acids
- Activates CD40 receptor on antigen-presenting cells
These proteins are stimulating B cell proliferation, immunoglobulin class switching, and antibody secretion. In peripheral blood monocytes, these are helps to induces cytokine production and tumoricidal activity in peripheral blood monocytes. These CD40L are also act as a co-stimulator for the activation of T cell proliferation and inflammatory protein production.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: Activity of HEK-blue™ CD40L cells, ED50 ≤50 ng/ml, Bioactivity ≥2,0×10⁴ units/mg
- Gene ID:P29965
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:TRAP|gp39|CD154|T-BAM|TNFSF5
- Protein/peptide name:CD40-Ligand
- Purity:≥95%
- Molecular weight:16,3 kDa
- Sequence:MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay