
Osmium(IV) oxide (≥83% Os)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12036-02-1
:
Formula:O₂Os
MDL Number:MFCD00011150
:222.23 g/mol
Density:11,4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together