

Cobalt(II) iodide, anhydrous ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15238-00-3
:
Formula:CoI₂
MDL Number:MFCD00016030
Merck Index:14,2443
Boiling Pt:570 °C (in vacuum)
Melting Pt:515 °C vac,
Density:5,68 g/cm³ (20 °C)
:312.74 g/mol
Storage Temperature:Ambient
EINECS:239-283-2
:GHS08
:GHS07
Related Information
Frequently Bought Together