


Platinum(IV) chloride (57,5% Pt), anhydrous for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13454-96-1
:
Formula:Cl₄Pt
MDL Number:MFCD00011182
Melting Pt:370 °C
Density:4,3 g/cm³ (20 °C)
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
:336.89 g/mol
EINECS:236-645-1
UN:2923
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:587 g/l (25 °C)
- Molar Mass:336.89 g/mol
- Density:4.3 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
Specification Test Results
Frequently Bought Together