


Platinum(IV) chloride ≥99.99% (metals basis) Pt ≥ 57%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13454-96-1
:
Formula:Cl₄Pt
MDL Number:MFCD00011182
Melting Pt:370 °C (dec.)
Density:4,303
Storage Temperature:Ambient
EINECS:236-645-1
UN:2923
ADR:8,III
:336.89 g/mol
:GHS06
:GHS08
:GHS05
Related Information
Frequently Bought Together