Order Entry
Northern Ireland
ContactUsLinkComponent
Platinum(II) chloride (≥73% Pt)
Health
Corrosive
Platinum(II) chloride (≥73% Pt)
Certificates
undefined
Health
Corrosive
Platinum(II) chloride (≥73% Pt)

Specification Test Results

  • Appearance:
    Grey-green to brown Powder
  • Assay:
    ≥71.8 % (Pt)

Specification Test Results

Related Information

Frequently Bought Together