Order Entry
Northern Ireland
ContactUsLinkComponent
Iridium(IV) chloride (≥56,5% Ir) ≥99.95% (metals basis), Premion®
Irritant
Iridium(IV) chloride (≥56,5% Ir) ≥99.95% (metals basis), Premion®
Catalog # 12184.06
CAS Number:  
Irritant
Iridium(IV) chloride (≥56,5% Ir) ≥99.95% (metals basis), Premion®
Catalog # 12184.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt: Dec.
 :334.03 g/mol
EINECS:233-048-8
 :GHS07
Storage Temperature:Ambient
CAS Number:10025-97-5
Formula:Cl₄Ir
MDL Number:MFCD00016155