
Platinum(IV) bromide (≥37,1% Pt) ≥99.99% (metals basis)
Catalog # 12171.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:180 °C (dec.)
Storage Temperature:Ambient
EINECS:273-151-5
ADR:8,III
:514.7 g/mol
UN:3260
:GHS05
CAS Number:68938-92-1
Formula:Br₄Pt
MDL Number:MFCD00016289