Supelco® Organic Reference Standard, Astm® D6584 1,2,4-Butanetriol Solution
Catalog # 44896-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:42890-76-6
- Description:ASTM® D6584 1,2,4-Butanetriol Solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H315 - H319
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Biodiesel impurity; Butanetriol solution
- Formula:C₄H₁₀O₃
- Storage temperature:2...8 °C
- Flash point:17 °C
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338