
Tungsten trioxide ≥99.998% (metals basis excluding Mo), Puratronic® (max. 0.01% Mo)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-35-8
:
Formula:O₃W
MDL Number:MFCD00011466
:231.84 g/mol
Merck Index:14,9817
Melting Pt:1473 °C
Density:7,16
EINECS:215-231-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together