

Tungsten(VI) chloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13283-01-7
:
Formula:Cl₆W
MDL Number:MFCD00011463
Boiling Pt:346 °C
Melting Pt:275 °C
Density:3,52
EINECS:236-293-9
UN:3260
ADR:8,II
:396.56 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together